Product Details
- SNP ID
-
rs3741699
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:114671444 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTTTTTTTTTGAAAGATTTTGTT[G/T]TTGTTTCCACTTGACACAGTGAAGG
- Phenotype
-
MIM: 601621
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TBX3
PubMed Links
Gene Details
- Gene
- TBX3
- Gene Name
- T-box 3
View Full Product Details