Product Details
- SNP ID
-
rs4650035
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
27
- Location
-
Chr.1:70268176 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTCACTCATACAGCTAGCTTCCCC[C/G]CCCCCACCTTGAGACAGAGTCTTGC
- Phenotype
-
MIM: 615125
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD13C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142997780] are located under a probe and SNP(s) [rs368839551] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKRD13C
- Gene Name
- ankyrin repeat domain 13C
View Full Product Details