Product Details

SNP ID
rs2758715
Assay Type
DME
NCBI dbSNP Submissions
3
Location
Chr.1:47142199 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TCCACAATGACATCCTGAAGCCATA[C/T]GTGGGGCTCATGGCAGACTCTGTAC
Phenotype
MIM: 615341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP4A22 PubMed Links

Gene Details

Gene
CYP4A22
Gene Name
cytochrome P450 family 4 subfamily A member 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010969.3 Intron NP_001010969.2
NM_001308102.1 Intron NP_001295031.1
XM_005270767.3 Intron XP_005270824.1
XM_005270770.3 Intron XP_005270827.1

View Full Product Details