Product Details

SNP ID
rs3176323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36679072 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATTACAAGTACAGGAATCCCTGG[C/T]CACGCTCCCCGCCCCTGGAAACCCA
Phenotype
MIM: 116899
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKN1A PubMed Links

Gene Details

Gene
CDKN1A
Gene Name
cyclin dependent kinase inhibitor 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000389.4 Intron NP_000380.1
NM_001220777.1 Intron NP_001207706.1
NM_001220778.1 Intron NP_001207707.1
NM_001291549.1 Intron NP_001278478.1
NM_078467.2 Intron NP_510867.1
Gene
PANDAR
Gene Name
promoter of CDKN1A antisense DNA damage activated RNA
There are no transcripts associated with this gene.

View Full Product Details