Product Details

SNP ID
rs4396031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:86150147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATACTCAAGATTATCTGACTTT[A/T]TAAACGCCTGTGAATTAGTCTTCTA
Phenotype
MIM: 300608
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DACH2 PubMed Links

Gene Details

Gene
DACH2
Gene Name
dachshund family transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139514.1 Intron NP_001132986.1
NM_001139515.1 Intron NP_001132987.1
NM_053281.3 Intron NP_444511.1

View Full Product Details