Product Details

SNP ID
rs4646295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69776994 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGTCCAAAGAAATACGTTGCTTT[G/T]ATTTAGTGTTTCCACTGGGGTCTTC
Phenotype
MIM: 182396 MIM: 600914
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100289511 PubMed Links

Gene Details

Gene
LOC100289511
Gene Name
uncharacterized LOC100289511
There are no transcripts associated with this gene.

Gene
SLC10A1
Gene Name
solute carrier family 10 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003049.3 Intron NP_003040.1
Gene
SRSF5
Gene Name
serine and arginine rich splicing factor 5
There are no transcripts associated with this gene.

View Full Product Details