Product Details

SNP ID
hCV32358054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:22172123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAACGGGACGTACCACCACCAC[A/C]ACCACCACCACCACCACCATCCGAG
Phenotype
MIM: 601656
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GATA6 PubMed Links

Gene Details

Gene
GATA6
Gene Name
GATA binding protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005257.5 1262 Missense Mutation AAC,CAC N327H NP_005248.2
Gene
GATA6-AS1
Gene Name
GATA6 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details