Product Details

SNP ID
rs4803750
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44744370 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCATGTTGAATTCTATTTTCCCCA[A/G]TGAAGCAGGTCCTGCTATTGAAAAC
Phenotype
MIM: 109560
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BCL3 PubMed Links

Gene Details

Gene
BCL3
Gene Name
B-cell CLL/lymphoma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005178.4 Intron NP_005169.2
XM_011527197.2 Intron XP_011525499.2
XM_011527198.2 Intron XP_011525500.2
XM_011527200.2 Intron XP_011525502.1
XM_017027109.1 Intron XP_016882598.1
XM_017027110.1 Intron XP_016882599.1

View Full Product Details