Product Details
- SNP ID
-
rs4505265
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:81185114 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTGGGGCGTAGGGTTTGAGGAGG[A/C]TTCTGCTGCACACCTTCCCCCACCA
- Phenotype
-
MIM: 603035
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL16
PubMed Links
Gene Details
- Gene
- IL16
- Gene Name
- interleukin 16
View Full Product Details