Product Details

SNP ID
rs3742178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:111305848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGTCTTTGATACTGAAGATAAC[A/G]TGTTTCCTTTAAAGTCTAGAGTTTG
Phenotype
MIM: 605477
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF7 PubMed Links

Gene Details

Gene
ARHGEF7
Gene Name
Rho guanine nucleotide exchange factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113511.2 11 Intron NP_001106983.1
NM_001113512.2 11 Intron NP_001106984.1
NM_001113513.1 11 Intron NP_001106985.1
NM_001320851.1 11 Intron NP_001307780.1
NM_001320852.1 11 Intron NP_001307781.1
NM_001320853.1 11 Intron NP_001307782.1
NM_001320854.1 11 Intron NP_001307783.1
NM_003899.3 11 Intron NP_003890.1
NM_145735.3 11 Intron NP_663788.1
XM_006719956.3 11 Intron XP_006720019.1
XM_011521133.2 11 Intron XP_011519435.1
XM_017020814.1 11 Intron XP_016876303.1
XM_017020815.1 11 Intron XP_016876304.1
XM_017020816.1 11 Intron XP_016876305.1
XM_017020817.1 11 Intron XP_016876306.1
XM_017020818.1 11 Intron XP_016876307.1
XM_017020819.1 11 Intron XP_016876308.1
XM_017020820.1 11 Intron XP_016876309.1
XM_017020821.1 11 Intron XP_016876310.1
XM_017020822.1 11 Intron XP_016876311.1
XM_017020823.1 11 Intron XP_016876312.1
XM_017020824.1 11 Intron XP_016876313.1
XM_017020825.1 11 Intron XP_016876314.1
XM_017020826.1 11 Intron XP_016876315.1
XM_017020827.1 11 Intron XP_016876316.1
XM_017020828.1 11 Intron XP_016876317.1
XM_017020829.1 11 Intron XP_016876318.1
Gene
TEX29
Gene Name
testis expressed 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303133.1 11 Intron NP_001290062.1
NM_152324.2 11 Intron NP_689537.1
XM_017020387.1 11 Intron XP_016875876.1
XM_017020388.1 11 UTR 5 XP_016875877.1

View Full Product Details