Product Details

SNP ID
rs4790126
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3289047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGAATTATACTATGCAATATAG[A/G]CAGAGAAAAGTCAGATTACAACACA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR3A1 PubMed Links

Gene Details

Gene
OR3A1
Gene Name
olfactory receptor family 3 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002550.2 Intron NP_002541.2
Gene
OR3A2
Gene Name
olfactory receptor family 3 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002551.3 Intron NP_002542.3

View Full Product Details