Product Details

SNP ID
rs16958241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68652365 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTTCCTCATATGCCACTGTGTT[C/T]CTAGGTTACCCAGTGCAGGGTGCTC
Phenotype
MIM: 114021
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH3 PubMed Links

Gene Details

Gene
CDH3
Gene Name
cadherin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317195.1 Intron NP_001304124.1
NM_001317196.1 Intron NP_001304125.1
NM_001793.5 Intron NP_001784.2
XM_011522800.2 Intron XP_011521102.1

View Full Product Details