Product Details

SNP ID
rs16938550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:73014889 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATAGAACTAGATAGAGGCAGAATA[A/G]TTGTGAAAGAGACAGAGCCATAGGT
Phenotype
MIM: 600951
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TERF1 PubMed Links

Gene Details

Gene
TERF1
Gene Name
telomeric repeat binding factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003218.3 Intron NP_003209.2
NM_017489.2 Intron NP_059523.2
XM_005251291.3 Intron XP_005251348.1
XM_005251292.3 Intron XP_005251349.1
XM_011517582.2 Intron XP_011515884.1
XM_017013792.1 Intron XP_016869281.1

View Full Product Details