Product Details

SNP ID
rs17110736
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:93997213 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAAAACCACACATGGAATGCAC[A/G]TTTATCAAAACTCACTAAACTGTAC
Phenotype
MIM: 601691
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA4 PubMed Links

Gene Details

Gene
ABCA4
Gene Name
ATP binding cassette subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000350.2 Intron NP_000341.2

View Full Product Details