Product Details

SNP ID
rs17116357
Assay Type
Functionally Tested
NCBI dbSNP Submissions
26
Location
Chr.1:97079204 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACCACAAAGGTCAACAATGTCCC[C/G]ATTTTAGCGTTAACATTTTTCTCTG
Phenotype
MIM: 612779
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
DPYD PubMed Links

Gene Details

Gene
DPYD
Gene Name
dihydropyrimidine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000110.3 Intron NP_000101.2
NM_001160301.1 Intron NP_001153773.1
XM_005270562.3 Intron XP_005270619.2
XM_006710397.3 Intron XP_006710460.1
XM_017000507.1 Intron XP_016855996.1
XM_017000508.1 Intron XP_016855997.1
XM_017000509.1 Intron XP_016855998.1
XM_017000510.1 Intron XP_016855999.1

View Full Product Details