Product Details

SNP ID
rs17287051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88669888 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCAGCCACAATCGACTTCATTGT[A/G]AAGAAAACTGGACAGAAGCAGCTAC
Phenotype
MIM: 601980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIPF PubMed Links

Gene Details

Gene
LIPF
Gene Name
lipase F, gastric type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198828.1 530 Silent Mutation GTA,GTG V125V NP_001185757.1
NM_001198829.1 530 Silent Mutation GTA,GTG V168V NP_001185758.1
NM_001198830.1 530 Silent Mutation GTA,GTG V135V NP_001185759.1
NM_004190.3 530 Silent Mutation GTA,GTG V158V NP_004181.1
XM_011540311.1 530 Silent Mutation GTA,GTG V158V XP_011538613.1

View Full Product Details