Product Details

SNP ID
rs17111189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88664680 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTTTTTTTGTATGGACATGAGA[C/T]TCTTAATTATGGAATGACAAATTTG
Phenotype
MIM: 601980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIPF PubMed Links

Gene Details

Gene
LIPF
Gene Name
lipase F, gastric type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198828.1 Intron NP_001185757.1
NM_001198829.1 Intron NP_001185758.1
NM_001198830.1 Intron NP_001185759.1
NM_004190.3 Intron NP_004181.1
XM_011540311.1 Intron XP_011538613.1

View Full Product Details