Product Details

SNP ID
rs17109503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88590153 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGAGTTGCTGAATGAAAGTTAT[C/G]CAAGCTTCCAAATCTCCCCGTTCTT
Phenotype
MIM: 613921 MIM: 609360
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LIPJ PubMed Links

Gene Details

Gene
LIPJ
Gene Name
lipase family member J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010939.2 Intron NP_001010939.2
XM_005269542.3 Intron XP_005269599.1
XM_006717635.2 Intron XP_006717698.1
XM_011539314.2 Intron XP_011537616.1
XM_011539315.2 Intron XP_011537617.1
XM_011539317.2 Intron XP_011537619.1
XM_011539318.2 Intron XP_011537620.1
XM_011539319.1 Intron XP_011537621.1
XM_011539320.2 Intron XP_011537622.1
XM_017015740.1 Intron XP_016871229.1
Gene
RNLS
Gene Name
renalase, FAD dependent amine oxidase
There are no transcripts associated with this gene.

View Full Product Details