Product Details

SNP ID
rs17000647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:75556520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTTATTATGGTTACAAAGCTAA[A/C]AATTCCACTTTGAATTACAGCACTG
Phenotype
MIM: 614829 MIM: 612535
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C4orf26 PubMed Links

Gene Details

Gene
C4orf26
Gene Name
chromosome 4 open reading frame 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206981.1 Intron NP_001193910.1
NM_001257072.1 Intron NP_001244001.1
NM_178497.3 Intron NP_848592.2
XM_011531668.2 Intron XP_011529970.1
Gene
THAP6
Gene Name
THAP domain containing 6
There are no transcripts associated with this gene.

View Full Product Details