Product Details

SNP ID
rs17016074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:89726127 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAACACAAGTGAATAAAACACATC[A/G]CAAAATGGTAAAATTTCATATTTAG
Phenotype
MIM: 163890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNCA PubMed Links

Gene Details

Gene
SNCA
Gene Name
synuclein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000345.3 994 UTR 3 NP_000336.1
NM_001146054.1 994 UTR 3 NP_001139526.1
NM_001146055.1 994 UTR 3 NP_001139527.1
NM_007308.2 994 UTR 3 NP_009292.1
XM_011532203.1 994 Intron XP_011530505.1
XM_011532204.2 994 Intron XP_011530506.1
XM_011532205.2 994 Intron XP_011530507.1
XM_011532206.1 994 Intron XP_011530508.1
XM_011532207.1 994 Intron XP_011530509.1
XM_011532208.2 994 UTR 3 XP_011530510.1
XM_017008562.1 994 UTR 3 XP_016864051.1
XM_017008563.1 994 UTR 3 XP_016864052.1

View Full Product Details