Product Details

SNP ID
rs17031753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43887775 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGGATCACTTTTATTTCAAACAA[C/T]TAAATACAAACCAATATTTTACCCC
Phenotype
MIM: 605460 MIM: 607544
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCG8 PubMed Links

Gene Details

Gene
ABCG8
Gene Name
ATP binding cassette subfamily G member 8
There are no transcripts associated with this gene.

Gene
LRPPRC
Gene Name
leucine rich pentatricopeptide repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133259.3 4977 UTR 3 NP_573566.2
XM_006711915.2 4977 UTR 3 XP_006711978.1
XM_006711916.3 4977 Intron XP_006711979.1
XM_011532473.1 4977 UTR 3 XP_011530775.1
XM_017003117.1 4977 UTR 3 XP_016858606.1

View Full Product Details