Product Details

SNP ID
rs17022762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99551796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTATATCCAAATAAATAAATACA[A/T]GATATCTGCCATTTCTAACTCAAAT
Phenotype
MIM: 601464
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AFF3 PubMed Links

Gene Details

Gene
AFF3
Gene Name
AF4/FMR2 family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025108.1 Intron NP_001020279.1
NM_002285.2 Intron NP_002276.2
XM_005263943.3 Intron XP_005264000.2
XM_005263945.3 Intron XP_005264002.1
XM_011511169.2 Intron XP_011509471.1
XM_011511170.2 Intron XP_011509472.1
XM_011511171.2 Intron XP_011509473.1
XM_011511172.2 Intron XP_011509474.1
XM_011511173.2 Intron XP_011509475.1
XM_011511174.2 Intron XP_011509476.1
XM_011511175.2 Intron XP_011509477.1
XM_011511176.2 Intron XP_011509478.1
XM_011511177.2 Intron XP_011509479.1
XM_011511178.2 Intron XP_011509480.1
XM_011511179.2 Intron XP_011509481.1
XM_017004085.1 Intron XP_016859574.1
XM_017004086.1 Intron XP_016859575.1
XM_017004087.1 Intron XP_016859576.1

View Full Product Details