Product Details

SNP ID
rs16859222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:46245812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATAAACCTGTAAATTGTTCTTTA[C/T]TATTGGCAATATTAGGTAATGTTAT
Phenotype
MIM: 137140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRA2 PubMed Links

Gene Details

Gene
GABRA2
Gene Name
gamma-aminobutyric acid type A receptor alpha2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005248080.3 6017 UTR 3 XP_005248137.1
XM_011513675.1 6017 UTR 3 XP_011511977.1
XM_011513676.1 6017 UTR 3 XP_011511978.1
XM_017007982.1 6017 UTR 3 XP_016863471.1
XM_017007983.1 6017 UTR 3 XP_016863472.1

View Full Product Details