Product Details

SNP ID
rs16949083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65895693 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACCCTCTGCCTTTCCCTTCTCCT[C/G]TGTATTGCCTGGTTTGACTTGTATT
Phenotype
MIM: 612454 MIM: 605570
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MEGF11 PubMed Links

Gene Details

Gene
MEGF11
Gene Name
multiple EGF like domains 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032445.2 5946 UTR 3 NP_115821.2
XM_017022670.1 5946 UTR 3 XP_016878159.1
XM_017022671.1 5946 UTR 3 XP_016878160.1
XM_017022672.1 5946 UTR 3 XP_016878161.1
XM_017022673.1 5946 Intron XP_016878162.1
XM_017022674.1 5946 UTR 3 XP_016878163.1
XM_017022675.1 5946 UTR 3 XP_016878164.1
Gene
RAB11A
Gene Name
RAB11A, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details