Product Details
- SNP ID
-
rs17044576
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
53
- Location
-
Chr.1:207461325 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACCACTAATTTTTACTTGTAAGGG[C/T]GAAGGAGATGGAACAATTCTGAGGA
- Phenotype
-
MIM: 120650
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74857976] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CR2
- Gene Name
- complement component 3d receptor 2
View Full Product Details