Product Details

SNP ID
rs16976875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:11983015 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATAGTGCTTTACCTTGGTGCAAAT[C/T]CTCTAATCCTGTTTAAAAATTAATC
Phenotype
MIM: 605922
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IMPA2 PubMed Links

Gene Details

Gene
IMPA2
Gene Name
inositol monophosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014214.2 Intron NP_055029.1
XM_011525659.2 Intron XP_011523961.1
XM_011525661.2 Intron XP_011523963.1

View Full Product Details