Product Details

SNP ID
rs17852067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:192144109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTCCAATTTCATGTAGCGAT[G/T]GTTCTCTGTACATACACACTGAAAG
Phenotype
MIM: 601513
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FGF12 PubMed Links

Gene Details

Gene
FGF12
Gene Name
fibroblast growth factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004113.5 543 Missense Mutation CAA,CCA Q149P NP_004104.3
NM_021032.4 543 Missense Mutation CAA,CCA Q211P NP_066360.1
XM_005247227.2 543 Missense Mutation CAA,CCA Q175P XP_005247284.1
XM_006713538.3 543 Missense Mutation CAA,CCA Q146P XP_006713601.1
XM_017005879.1 543 Missense Mutation CAA,CCA Q125P XP_016861368.1

View Full Product Details