Product Details

SNP ID
rs17003000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23639999 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGGGACTGTCTCTAAGGCCACC[A/G]TACGGCCCCTCATCAGAACCGGCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRICH1 PubMed Links

Gene Details

Gene
DRICH1
Gene Name
aspartate rich 1
There are no transcripts associated with this gene.

Gene
GUSBP11
Gene Name
glucuronidase, beta pseudogene 11
There are no transcripts associated with this gene.

View Full Product Details