Product Details

SNP ID
rs16970453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20624199 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACAGGAACTGTGGGGTCAGGACA[A/G]TAAAGGCCTTCACCACCTGCAGAAT
Phenotype
MIM: 614357
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSM1 PubMed Links

Gene Details

Gene
ACSM1
Gene Name
acyl-CoA synthetase medium-chain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318890.1 2439 Missense Mutation ACT,ATT T515I NP_001305819.1
NM_052956.2 2439 Missense Mutation ACT,ATT T515I NP_443188.2
XM_006721016.2 2439 Missense Mutation ACT,ATT T481I XP_006721079.1
XM_006721017.2 2439 Missense Mutation ACT,ATT T454I XP_006721080.1
XM_006721018.2 2439 Missense Mutation ACT,ATT T295I XP_006721081.1
XM_011545729.2 2439 Intron XP_011544031.1
XM_011545730.2 2439 Missense Mutation ACT,ATT T357I XP_011544032.1
XM_011545731.2 2439 Missense Mutation ACT,ATT T316I XP_011544033.1
XM_011545732.2 2439 Missense Mutation ACT,ATT T295I XP_011544034.1
XM_017022914.1 2439 Intron XP_016878403.1
XM_017022915.1 2439 Missense Mutation ACT,ATT T277I XP_016878404.1

View Full Product Details