Product Details

SNP ID
rs16861748
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:162396277 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAATTTGGTATATTTCAAACTGA[A/C]CAGGTCCTAATTCACATTTTGAACT
Phenotype
MIM: 608510
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf226 PubMed Links

Gene Details

Gene
C1orf226
Gene Name
chromosome 1 open reading frame 226
There are no transcripts associated with this gene.

Gene
SH2D1B
Gene Name
SH2 domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053282.4 1524 UTR 3 NP_444512.2

View Full Product Details