Product Details

SNP ID
rs17009505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:30452470 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAGGCCGTCACATCATGCTGTTT[G/T]ACAATTTGGTCACAAAATTATGCCC
Phenotype
MIM: 614241
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LCLAT1 PubMed Links

Gene Details

Gene
LCLAT1
Gene Name
lysocardiolipin acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002257.2 Intron NP_001002257.1
NM_001304445.1 Intron NP_001291374.1
NM_001304446.1 Intron NP_001291375.1
NM_182551.4 Intron NP_872357.2
XM_005264244.1 Intron XP_005264301.1
XM_005264245.3 Intron XP_005264302.1
XM_011532741.2 Intron XP_011531043.1
XM_011532742.2 Intron XP_011531044.1
XM_011532743.2 Intron XP_011531045.1
XM_011532744.2 Intron XP_011531046.1
XM_011532745.1 Intron XP_011531047.1
XM_017003746.1 Intron XP_016859235.1
XM_017003747.1 Intron XP_016859236.1

View Full Product Details