Product Details

SNP ID
rs16857265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:11190286 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATATGAGGGAAAGAAATCCTTAC[C/T]ACTTACCGTTGCTATGACAAAATGT
Phenotype
MIM: 604002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ROCK2 PubMed Links

Gene Details

Gene
ROCK2
Gene Name
Rho associated coiled-coil containing protein kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321643.1 Intron NP_001308572.1
NM_004850.4 Intron NP_004841.2
XM_005246190.3 Intron XP_005246247.1
XM_011510417.2 Intron XP_011508719.1
XM_017005378.1 Intron XP_016860867.1
XM_017005379.1 Intron XP_016860868.1

View Full Product Details