Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001009877.2 | 2318 | UTR 3 | NP_001009877.2 | ||
NM_001317951.1 | 2318 | UTR 3 | NP_001304880.1 | ||
NM_023924.4 | 2318 | UTR 3 | NP_076413.3 | ||
XM_006714494.3 | 2318 | Intron | XP_006714557.1 | ||
XM_017009780.1 | 2318 | Intron | XP_016865269.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_024786.2 | 2318 | Intron | NP_079062.1 | ||
XM_017009869.1 | 2318 | Intron | XP_016865358.1 | ||
XM_017009870.1 | 2318 | Intron | XP_016865359.1 | ||
XM_017009871.1 | 2318 | Intron | XP_016865360.1 | ||
XM_017009872.1 | 2318 | Intron | XP_016865361.1 | ||
XM_017009873.1 | 2318 | Intron | XP_016865362.1 | ||
XM_017009874.1 | 2318 | Intron | XP_016865363.1 | ||
XM_017009875.1 | 2318 | Intron | XP_016865364.1 | ||
XM_017009876.1 | 2318 | Intron | XP_016865365.1 | ||
XM_017009877.1 | 2318 | Intron | XP_016865366.1 | ||
XM_017009878.1 | 2318 | Intron | XP_016865367.1 |