Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_144980.3 | 1317 | Missense Mutation | NP_659417.2 | ||
XM_005266838.2 | 1317 | Missense Mutation | XP_005266895.1 | ||
XM_011535509.2 | 1317 | Missense Mutation | XP_011533811.1 | ||
XM_011535510.2 | 1317 | Missense Mutation | XP_011533812.1 | ||
XM_011535511.2 | 1317 | Missense Mutation | XP_011533813.1 | ||
XM_011535512.2 | 1317 | Missense Mutation | XP_011533814.1 | ||
XM_011535513.2 | 1317 | Missense Mutation | XP_011533815.1 | ||
XM_017010323.1 | 1317 | Missense Mutation | XP_016865812.1 | ||
XM_017010324.1 | 1317 | Missense Mutation | XP_016865813.1 |