Product Details
- SNP ID
-
rs17281535
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:119173624 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGAGCGGGCCCTACCGTGTGCGCA[G/T]AAAGAGGAGGCGCTGTGAGTAGACC
- Phenotype
-
MIM: 602380
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
UPK1B
PubMed Links
Gene Details
- Gene
- UPK1B
- Gene Name
- uroplakin 1B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006952.3 |
47 |
UTR 5 |
|
|
NP_008883.2 |
View Full Product Details