Product Details

SNP ID
rs17041429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:15671364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAATAGTGTGATGCCAAGCTTCA[A/G]TTATACCATATGGAACTTTAATAGT
Phenotype
MIM: 611122
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD28 PubMed Links

Gene Details

Gene
ANKRD28
Gene Name
ankyrin repeat domain 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195098.1 Intron NP_001182027.1
NM_001195099.1 Intron NP_001182028.1
NM_015199.3 Intron NP_056014.2
XM_005264996.3 Intron XP_005265053.1
XM_011533540.2 Intron XP_011531842.1
XM_011533541.2 Intron XP_011531843.1
XM_011533542.2 Intron XP_011531844.1
XM_011533543.2 Intron XP_011531845.1
XM_011533545.2 Intron XP_011531847.1
XM_011533546.2 Intron XP_011531848.1
XM_011533547.2 Intron XP_011531849.1
XM_017006026.1 Intron XP_016861515.1
XM_017006027.1 Intron XP_016861516.1
XM_017006028.1 Intron XP_016861517.1
XM_017006029.1 Intron XP_016861518.1

View Full Product Details