Product Details

SNP ID
rs17163344
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:25884984 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCCTAAAATCCCTTTGGTATCTA[C/T]AGGAGGAGGTTGTGAAGTCTGTAAG
Phenotype
MIM: 607779 MIM: 151442
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAQR7 PubMed Links

Gene Details

Gene
PAQR7
Gene Name
progestin and adipoQ receptor family member 7
There are no transcripts associated with this gene.

Gene
STMN1
Gene Name
stathmin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145454.2 1440 UTR 3 NP_001138926.1
NM_005563.3 1440 Intron NP_005554.1
NM_203399.1 1440 Intron NP_981944.1
NM_203401.1 1440 Intron NP_981946.1

View Full Product Details