Product Details

SNP ID
rs17654140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122993494 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATGGACCATAAATGTGAGAAAT[C/T]CAAATGAACATCTTGTAAGATTATG
Phenotype
MIM: 606238 MIM: 611310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IKZF5 PubMed Links

Gene Details

Gene
IKZF5
Gene Name
IKAROS family zinc finger 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271840.1 2345 UTR 3 NP_001258769.1
XM_006717947.3 2345 UTR 3 XP_006718010.1
XM_017016550.1 2345 UTR 3 XP_016872039.1
XM_017016551.1 2345 UTR 3 XP_016872040.1
XM_017016552.1 2345 UTR 3 XP_016872041.1
XM_017016553.1 2345 UTR 3 XP_016872042.1
XM_017016554.1 2345 UTR 3 XP_016872043.1
XM_017016555.1 2345 UTR 3 XP_016872044.1
Gene
PSTK
Gene Name
phosphoseryl-tRNA kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153336.2 2345 Intron NP_699167.2
XM_017015640.1 2345 Intron XP_016871129.1
XM_017015641.1 2345 Intron XP_016871130.1

View Full Product Details