Product Details

SNP ID
rs17436125
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:183632196 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGATGTATGAATAGCTATGGACAA[C/T]GGGGTTGATGCTATGGGCATCATTT
Phenotype
MIM: 604227 MIM: 605667
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC5 PubMed Links

Gene Details

Gene
ARPC5
Gene Name
actin related protein 2/3 complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270439.1 Intron NP_001257368.1
NM_005717.3 Intron NP_005708.1
Gene
RGL1
Gene Name
ral guanine nucleotide dissociation stimulator like 1
There are no transcripts associated with this gene.

View Full Product Details