Product Details
- SNP ID
-
rs17852944
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:102813475 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCATAGCCAAACCAACGTGGAGGG[C/T]CATTAGTGTTGTATTCCTGCTGCTG
- Phenotype
-
MIM: 609080
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM185A
PubMed Links
Gene Details
- Gene
- FAM185A
- Gene Name
- family with sequence similarity 185 member A
- Gene
- FBXL13
- Gene Name
- F-box and leucine rich repeat protein 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001111038.1 |
2306 |
Missense Mutation |
GAC,GGC |
D647G |
NP_001104508.1 |
NM_001287150.1 |
2306 |
Missense Mutation |
GAC,GGC |
D664G |
NP_001274079.1 |
NM_145032.3 |
2306 |
Missense Mutation |
GAC,GGC |
D692G |
NP_659469.3 |
XM_005250205.3 |
2306 |
Missense Mutation |
GAC,GGC |
D782G |
XP_005250262.1 |
XM_005250207.3 |
2306 |
Missense Mutation |
GAC,GGC |
D749G |
XP_005250264.1 |
XM_005250208.3 |
2306 |
Missense Mutation |
GAC,GGC |
D737G |
XP_005250265.1 |
XM_005250209.2 |
2306 |
Missense Mutation |
GAC,GGC |
D692G |
XP_005250266.1 |
XM_006715898.2 |
2306 |
Missense Mutation |
GAC,GGC |
D541G |
XP_006715961.1 |
XM_011515928.2 |
2306 |
Missense Mutation |
GAC,GGC |
D754G |
XP_011514230.1 |
XM_011515929.2 |
2306 |
Intron |
|
|
XP_011514231.1 |
XM_011515930.2 |
2306 |
Missense Mutation |
GAC,GGC |
D692G |
XP_011514232.1 |
XM_011515932.2 |
2306 |
Intron |
|
|
XP_011514234.1 |
XM_017011850.1 |
2306 |
Missense Mutation |
GAC,GGC |
D779G |
XP_016867339.1 |
XM_017011851.1 |
2306 |
Intron |
|
|
XP_016867340.1 |
XM_017011852.1 |
2306 |
Missense Mutation |
GAC,GGC |
D737G |
XP_016867341.1 |
XM_017011853.1 |
2306 |
Missense Mutation |
GAC,GGC |
D692G |
XP_016867342.1 |
View Full Product Details