Product Details

SNP ID
rs17002150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40770679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATTAAGGTGACAACAGGTCCAG[G/T]TGGCCATACTCCCTGTGCACCCTTG
Phenotype
MIM: 606795
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A17 PubMed Links

Gene Details

Gene
SLC25A17
Gene Name
solute carrier family 25 member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282726.1 1283 UTR 3 NP_001269655.1
NM_001282727.1 1283 UTR 3 NP_001269656.1
NM_006358.3 1283 UTR 3 NP_006349.1

View Full Product Details