Product Details

SNP ID
rs16986475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:111299039 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTGCTGACTTGATGTTTGTTGAC[C/T]GTGCAGAAATTAACACAATGGAACT
Phenotype
MIM: 300121
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCX PubMed Links

Gene Details

Gene
DCX
Gene Name
doublecortin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000555.3 3931 UTR 3 NP_000546.2
NM_001195553.1 3931 UTR 3 NP_001182482.1
NM_178151.2 3931 UTR 3 NP_835364.1
NM_178152.2 3931 UTR 3 NP_835365.1
NM_178153.2 3931 UTR 3 NP_835366.1
XM_011530878.2 3931 UTR 3 XP_011529180.1
XM_011530879.2 3931 UTR 3 XP_011529181.1
XM_011530880.2 3931 UTR 3 XP_011529182.1
XM_017029312.1 3931 UTR 3 XP_016884801.1

View Full Product Details