Product Details

SNP ID
rs34069017
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:109075686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTGATCTCTGATGATTCTGTAC[A/G]TGTTTCAGTGTCTGGCAAATACAGA
Phenotype
MIM: 600774
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAF13 PubMed Links

Gene Details

Gene
TAF13
Gene Name
TATA-box binding protein associated factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005645.3 Intron NP_005636.1

View Full Product Details