Product Details

SNP ID
rs17855517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41494139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGACAGGTCCCCAACGGGCTTC[A/C]GCGGGCTCCCCCGGCCCCGGTGATC
Phenotype
MIM: 147521 MIM: 151520
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ITPKA PubMed Links

Gene Details

Gene
ITPKA
Gene Name
inositol-trisphosphate 3-kinase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002220.2 282 Missense Mutation CAG,CCG Q71P NP_002211.1
XM_011521522.2 282 Intron XP_011519824.1
Gene
LTK
Gene Name
leukocyte receptor tyrosine kinase
There are no transcripts associated with this gene.

View Full Product Details