Product Details

SNP ID
rs17800789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44510962 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTTCATCCTACAGACTCTTTAG[C/T]AGGGAAGAAAAAGCAGATTGGTGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEACAM20 PubMed Links

Gene Details

Gene
CEACAM20
Gene Name
carcinoembryonic antigen related cell adhesion molecule 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102597.2 Intron NP_001096067.2
NM_001102598.2 Intron NP_001096068.2
NM_001102599.2 Intron NP_001096069.2
NM_001102600.2 Intron NP_001096070.2
XM_011526429.2 Intron XP_011524731.1
XM_011526430.2 Intron XP_011524732.1
XM_011526431.2 Intron XP_011524733.1
XM_011526432.2 Intron XP_011524734.1

View Full Product Details