Product Details
- SNP ID
-
rs17714646
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:44538508 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGTCACAATGCAGTTGACATCTCC[A/C]TGCCTCTGGAAAATATGAAACCTTG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CEACAM20
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146241111] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEACAM20
- Gene Name
- carcinoembryonic antigen related cell adhesion molecule 20
There are no transcripts associated with this gene.
- Gene
- CEACAM22P
- Gene Name
- carcinoembryonic antigen related cell adhesion molecule 22, pseudogene
There are no transcripts associated with this gene.
View Full Product Details