Product Details

SNP ID
rs17342532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103275148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTAGGGTCTTGACAGCAGTCA[C/T]GTATCCCGAAACTGCAAGAGGGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCEAL5 PubMed Links

Gene Details

Gene
TCEAL5
Gene Name
transcription elongation factor A like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012979.2 Intron NP_001012997.1

View Full Product Details