Product Details
- SNP ID
-
rs17138512
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:37686595 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTACTCTGACACACTACATTCAGC[C/T]CCTCCAGGGATCCAGAGGTGTGTCT
- Phenotype
-
MIM: 189907
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HNF1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75361710] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HNF1B
- Gene Name
- HNF1 homeobox B
View Full Product Details