Product Details

SNP ID
rs72559707
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.10:133527505 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GCTGGAATCTGCCCCCAGGCCCTTT[-/C]CCCGCTTCCCATCATCGGGAACCTC
Phenotype
MIM: 124040
Polymorphism
-/C, Insertion/deletion
Allele Nomenclature
Literature Links
CYP2E1 PubMed Links
Additional Information
The CYP2E1 gene exhibits copy number variation. Individuals may carry extra copies of CYP2E1. CYP2E1 SNP genotyping assays run on samples having 2 or more gene copies that are homozygous for the SNP allele will cluster together, and samples having more than 2 gene copies that are heterozygous may run between the 2 copy heterozygous and homozygous clusters. For accurate CYP2E1 genotype analysis, copy number analysis must be done. For more information, refer to the PGx Experiments User Guide (Pub. # MAN0009612) Chapter 2 Copy Number Variation section.

Gene Details

Gene
CYP2E1
Gene Name
cytochrome P450 family 2 subfamily E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000773.3 143 Frame Shift InDel TCT,TTC S37F NP_000764.1

View Full Product Details