Product Details

SNP ID
rs17034260
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:115702529 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTGCCTGTAGAGTAGTTGCCTGC[C/T]TGGGATTCTAGGCCACAACGGCCCC
Phenotype
MIM: 114251 MIM: 610132
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASQ2 PubMed Links

Gene Details

Gene
CASQ2
Gene Name
calsequestrin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001232.3 Intron NP_001223.2
Gene
VANGL1
Gene Name
VANGL planar cell polarity protein 1
There are no transcripts associated with this gene.

View Full Product Details